Description
- Includes additional measures for the greatest possible impermeability to gas
- Not in conjunction with access port or interior lighting option
Specifications
Specifications
| Description | Gas-tight option model - Incl. additional measures for the greatest possible impermeability to gas; not in conjunction with access port or interior lighting option |
| For Use With (Equipment) | BINDER Chambers FED 115 |
| Material | Steel |
| Type | Gas-tight Option Model |