Learn More
Chemglass Life Sciences Gas Dispersion Tube, Large Capacity, Fritted, Extra Coarse Porosity
SDP
Catalog No. 50873746
Encompass_Preferred
4 Ring Glassblower Hose Connection. "A" = 2.5, "B" = 6.5, "C" = 8, "D" = 20; "E" = 8. Serrations are tooled on both ends of an 8" length of tubing. Hose connections are SOLD IN PACKAGES OF 10 SERRATED CONNECTORS (5 pieces tooled on both ends).
Catalog No. 50-873-746 Supplier Chemglass Life Sciences Supplier No. CG22002
4 Ring Glassblower Hose Connection. "A" = 2.5, "B" = 6.5, "C" = 8, "D" = 20; "E" = 8. Serrations are tooled on both ends of an 8" length of tubing. Hose connections are SOLD IN PACKAGES OF 10 SERRATED CONNECTORS (5 pieces tooled on both ends).