Learn More
Grainger Dormont Gas Tubing Kit, Stainless Steel, 1/4 in. x 30 in.
Catalog No. 502778596
Encompass_Preferred
This pilot flex tube kit is designed to replace damaged internal supply lines on commercial cooking equipment. It provides a robust, flexible alternative to rigid aluminum tubing, helping to minimize time spent on the job site by eliminating the need for flaring during assembly.
  • Features corrugated stainless steel construction
  • Designed for use with SAE J512 compression fittings
  • Operating temperature range of -40°F to +500°F
  • Working pressure rating of 0 to 15 PSIG
  • BTU capacity of 21,500
  • Includes one flex tube, two compression sleeves, two compression nuts, and one break-away fitting
  • Compatible with 1/4 in. applications
Catalog No. 50-277-8596 Supplier Grainger Supplier No. 18D849
This pilot flex tube kit is designed to replace damaged internal supply lines on commercial cooking equipment. It provides a robust, flexible alternative to rigid aluminum tubing, helping to minimize time spent on the job site by eliminating the need for flaring during assembly.
  • Features corrugated stainless steel construction
  • Designed for use with SAE J512 compression fittings
  • Operating temperature range of -40°F to +500°F
  • Working pressure rating of 0 to 15 PSIG
  • BTU capacity of 21,500
  • Includes one flex tube, two compression sleeves, two compression nuts, and one break-away fitting
  • Compatible with 1/4 in. applications