Learn More
Hach Company GAS WASHING BOTTLE, 1200ML
Supplier:  Hach Company 2662200
Encompass
For use in precipitating elements from gas. Used in the Hach Cold Vapor Mercury Apparatus Set (Prod. No. 2674400) to perform Cold Vapor Mercury Concentration Analysis–Hach Method 10065.
Catalog No. 50-199-6502