Learn More
Ampco Gas and Water Shut-off Wrench
SDP
Catalog No. 19174628 Shop All Ampco Metal Inc. Products
Change view
Click to view available options
Length (English):
11 to 1/2 in.
Catalog No. Length (English)
19-174-628 11 to 1/2 in.
1 options
Catalog No. 19-174-628 Supplier Ampco Supplier No. W20
Add to Cart
Add to Cart
Non-magnetic, non-sparking, and corrosion-resistant wrench. Made in the USA.
  • Please see image for more information regarding measurements.
  • Wrench size (s): 1/2 in.
  • Length: 11 and 1/2 in.
Specifications
Length (English) 11 to 1/2 in.
Length (Metric) 292.1 mm
Material Aluminum Bronze
No. per Pack 1 Ea.
Product Type Gas and Water Shut-Off Wrench