Learn More
Aviva Systems Biology Corp GAS2L1 Antibody - middle region (ARP52196_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP52196_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against GAS2L1. It was validated on Western Blot. . Alternate names: GAR22, MGC17243ApplicationWB. Species reactivity: Cow, Guinea Pig, Human, Mouse, Rat
Catalog No. 50-285-659