Learn More
Aviva Systems Biology Corp GAS7 Antibody - N-terminal region (ARP30004_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP30004_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against GAS7. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the N terminal region of human GAS7. Alternate names: MLL/GAS7ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-54093