Learn More
Aviva Systems Biology Corp GAS7 Antibody - N-terminal region (ARP47758_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP47758_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against GAS7. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human GAS7. Alternate names: KIAA0394, MGC1348, MLL/GAS7ApplicationWB. Species reactivity: Human, Pig
Catalog No. 50-014-59658