Learn More
ABclonal Technology GAS7 Rabbit pAb
SDP
Supplier:  ABclonal Technology A14179500UL
Encompass_Preferred
Growth arrest-specific 7 is expressed primarily in terminally differentiated brain cells and predominantly in mature cerebellar Purkinje neurons. GAS7 plays a putative role in neuronal development. Several transcript variants encoding proteins which vary in the N-terminus have been described.
Catalog No. 50-001-06952