Learn More
Aviva Systems Biology Corp GAS8 Antibody - middle region (ARP48017_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP48017_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against GAS8. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human GAS8. Alternate names: GAS11, MGC138326ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-53937