Learn More
MSC GASALERT DETECTOR
Catalog No. 5000686571
Encompass
The GasAlert Extreme is a durable and comfortable single gas detector, designed to reliably monitor for a specific gas hazard. It features easy on/off operation, extended longevity with a field-replaceable battery and sensor, and simple automatic calibration, making it suitable for personal safety in hazardous gas environments.
  • Direct, real-time readings on large LCD
  • Operates through consecutive shifts
  • Secure and accurate history with datalogging
  • Equipped with internal vibrating alarm for high noise areas
  • Compact and lightweight design
  • Simple automatic calibration procedure; compatible with MicroDock II automatic test and calibration station
Catalog No. 50-006-86571 Supplier MSC Supplier No. 46893400
The GasAlert Extreme is a durable and comfortable single gas detector, designed to reliably monitor for a specific gas hazard. It features easy on/off operation, extended longevity with a field-replaceable battery and sensor, and simple automatic calibration, making it suitable for personal safety in hazardous gas environments.
  • Direct, real-time readings on large LCD
  • Operates through consecutive shifts
  • Secure and accurate history with datalogging
  • Equipped with internal vibrating alarm for high noise areas
  • Compact and lightweight design
  • Simple automatic calibration procedure; compatible with MicroDock II automatic test and calibration station