Learn More
Cell Signaling Technology Gasdermin E Rabbit Monoclonal Antibody, Mouse, 100 μL
SDP
Catalog No. 502730352
Encompass
Gasdermin E (E1C5B) Rabbit Monoclonal Antibody is a high-quality reagent designed to detect endogenous levels of total Gasdermin E protein. This monoclonal antibody is also capable of identifying the amino-terminal cleavage fragment associated with pyroptosis, a lytic form of programmed cell death. It is produced by immunizing animals with a synthetic peptide corresponding to residues surrounding His112 of the mouse Gasdermin E protein, ensuring specific recognition.
  • Validated for western blotting applications.
  • Recognizes endogenous levels of total protein and the amino-terminal cleavage fragment.
  • Supplied in a stable formulation containing 10 mM sodium HEPES (pH 7.5), 150 mM NaCl, 100 μg/mL BSA, 50% glycerol, and <0.02% sodium azide.
  • Recommended storage at -20°C.
  • Do not aliquot the antibody.
Catalog No. 50-273-0352 Supplier Cell Signaling Technology Supplier No. 88874S
Gasdermin E (E1C5B) Rabbit Monoclonal Antibody is a high-quality reagent designed to detect endogenous levels of total Gasdermin E protein. This monoclonal antibody is also capable of identifying the amino-terminal cleavage fragment associated with pyroptosis, a lytic form of programmed cell death. It is produced by immunizing animals with a synthetic peptide corresponding to residues surrounding His112 of the mouse Gasdermin E protein, ensuring specific recognition.
  • Validated for western blotting applications.
  • Recognizes endogenous levels of total protein and the amino-terminal cleavage fragment.
  • Supplied in a stable formulation containing 10 mM sodium HEPES (pH 7.5), 150 mM NaCl, 100 μg/mL BSA, 50% glycerol, and <0.02% sodium azide.
  • Recommended storage at -20°C.
  • Do not aliquot the antibody.