Learn More
Grainger Sanitary Gaskets, Gaskets
Catalog No. NC2592585
Encompass_Preferred
The Grainger website features sanitary gaskets for use in food, beverage, pharmaceutical, and other sanitary process applications. They offer a variety of gasket materials including silicone, EPDM, Viton, PTFE, and more. Gaskets come in standard and custom shapes and sizes to fit pipes, vessels, pumps, valves, and other equipment. Key features include resistance to high temperatures, chemicals, and compression set. Grainger provides technical specs for each gasket to help customers select the optimal material, hardness, and design for their application. With a wide selection and technical expertise, Grainger is a leading supplier of sanitary gaskets for sanitary process industries.
Catalog No. NC2592585 Supplier Grainger Supplier No. 5PYD1
The Grainger website features sanitary gaskets for use in food, beverage, pharmaceutical, and other sanitary process applications. They offer a variety of gasket materials including silicone, EPDM, Viton, PTFE, and more. Gaskets come in standard and custom shapes and sizes to fit pipes, vessels, pumps, valves, and other equipment. Key features include resistance to high temperatures, chemicals, and compression set. Grainger provides technical specs for each gasket to help customers select the optimal material, hardness, and design for their application. With a wide selection and technical expertise, Grainger is a leading supplier of sanitary gaskets for sanitary process industries.