Learn More
Waters Corp Waters Corporation Probe Tip Gasket, 2 pk, MS Washer Type
Catalog No. 50792957 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This probe tip gasket is designed as a replacement part for original ESI probes found in early Xevo sourced Mass Spectrometers. It is also compatible with later instruments equipped with standard tool-free ESI probes. Each package contains 2 gaskets.
  • Replacement probe tip gasket for ESI probes.
  • Designed for original ESI probes on early mass spectrometers.
  • Compatible with later instruments featuring tool-free ESI probes.
  • Supplied in a 2 pack.
  • Washer type.
  • Suitable for Mass Spectrometry (MS) systems.
  • Replaced as required for maintenance.
Catalog No. 50-792-957 Supplier Waters Corp Supplier No. 700004208
This probe tip gasket is designed as a replacement part for original ESI probes found in early Xevo sourced Mass Spectrometers. It is also compatible with later instruments equipped with standard tool-free ESI probes. Each package contains 2 gaskets.
  • Replacement probe tip gasket for ESI probes.
  • Designed for original ESI probes on early mass spectrometers.
  • Compatible with later instruments featuring tool-free ESI probes.
  • Supplied in a 2 pack.
  • Washer type.
  • Suitable for Mass Spectrometry (MS) systems.
  • Replaced as required for maintenance.