Learn More
Chemglass Life Sciences Gasket, 400mm, Perfluoroelastomer, Centering Tabs, Class VI FFKM, -15°C to 275°C
SDP
Catalog No. 501531325
Encompass
Gasket, 400mm, Perfluoroelastomer, Centering Tabs, Class VI FFKM, -15C to 275C. Class VI Perfluoroelastomer with 3 tabs each spaced 120 degrees apart along the outer edge to aid in gasket alignment. The chemically resistant perfluoroelestomer has a soft pliable sealing surface that provides improved sealing under full vacuum, as opposed to encapsulated o-ring materials and envelope gaskets. Gasket is for use with 400mm Chemglass flat ground flange on Chemglass 50L and 100L vessels.
Catalog No. 50-153-1325 Supplier Chemglass Life Sciences Supplier No. CG1968H02
Gasket, 400mm, Perfluoroelastomer, Centering Tabs, Class VI FFKM, -15C to 275C. Class VI Perfluoroelastomer with 3 tabs each spaced 120 degrees apart along the outer edge to aid in gasket alignment. The chemically resistant perfluoroelestomer has a soft pliable sealing surface that provides improved sealing under full vacuum, as opposed to encapsulated o-ring materials and envelope gaskets. Gasket is for use with 400mm Chemglass flat ground flange on Chemglass 50L and 100L vessels.