Learn More
Grainger Gasket
Catalog No. 502766660
Encompass_Preferred
The Grainger website offers a wide selection of pipe fitting gaskets for various applications. These gaskets are made from materials like rubber, cork, fiber, PTFE, and more to provide a tight seal between pipe connections. They come in different sizes to fit standard pipe diameters. Common types include O-ring gaskets, flat gaskets, cone gaskets, and raised face gaskets. Factors to consider when selecting include temperature range, pressure rating, and chemical resistance. Grainger provides technical specifications for each gasket to help customers choose the right one for their needs. These gaskets are an important part of piping systems to prevent leaks.
Catalog No. 50-276-6660 Supplier Grainger Supplier No. 16X429
The Grainger website offers a wide selection of pipe fitting gaskets for various applications. These gaskets are made from materials like rubber, cork, fiber, PTFE, and more to provide a tight seal between pipe connections. They come in different sizes to fit standard pipe diameters. Common types include O-ring gaskets, flat gaskets, cone gaskets, and raised face gaskets. Factors to consider when selecting include temperature range, pressure rating, and chemical resistance. Grainger provides technical specifications for each gasket to help customers choose the right one for their needs. These gaskets are an important part of piping systems to prevent leaks.