Learn More
Cole-Parmer Argos Technologies Vortamix™ Mini Vortexer Gasket Cap, Red; 1/Ea
Catalog No. 501960668 Shop All Cole-Parmer Products
Encompass_Preferred
The standard unit comes with a red rubber gasket cap, but blue, green and purple colored caps are also available. Vortamix mini vortexer sold separately as model 04397-27. The innovative design of the gasket cap allows centrifuge and test tubes up to 30 mm in diameter to be mixed with very little effort – including 50mL conical tubes!
Catalog No. 50-196-0668 Supplier Cole-Parmer Supplier No. 0439729
The standard unit comes with a red rubber gasket cap, but blue, green and purple colored caps are also available. Vortamix mini vortexer sold separately as model 04397-27. The innovative design of the gasket cap allows centrifuge and test tubes up to 30 mm in diameter to be mixed with very little effort – including 50mL conical tubes!