Learn More
MSC CRC Gasket Remover, No Methylene Chloride, 12 oz
Catalog No. 502870067
Encompass
Removes baked on fiber gaskets, gasket adhesives and sealants. Dissolves and aids removal of paints, lacquers, adhesives, and varnish. Contains no methylene chloride.
  • Reduces the need for excessive scraping
  • Loosens carbon deposits
  • Contains no methylene chloride
  • NSF C3 Registered for use in meat and poultry plants
Catalog No. 50-287-0067 Supplier MSC Supplier No. 55310460
Removes baked on fiber gaskets, gasket adhesives and sealants. Dissolves and aids removal of paints, lacquers, adhesives, and varnish. Contains no methylene chloride.
  • Reduces the need for excessive scraping
  • Loosens carbon deposits
  • Contains no methylene chloride
  • NSF C3 Registered for use in meat and poultry plants