Learn More
Chemglass Life Sciences Gasket, Sanitary, 1", PTFE/Elastomer Core
SDP
Catalog No. 501311068
Encompass_Preferred
Gasket, Sanitary, 1", PTFE/Elastomer Core. Gasket grafts a PTFE envelope to an EPDM elastomer core. Gasket will not creep or cold flow overtime so it maintains the integrity of its seal. Temperature range -73 to 177¿ C.
Catalog No. 50-131-1068 Supplier Chemglass Life Sciences Supplier No. CG152G10
Gasket, Sanitary, 1", PTFE/Elastomer Core. Gasket grafts a PTFE envelope to an EPDM elastomer core. Gasket will not creep or cold flow overtime so it maintains the integrity of its seal. Temperature range -73 to 177¿ C.