Learn More
Chemglass Life Sciences Gasket, Sanitary, 2", PTFE/Elastomer Core
SDP
Catalog No. 501311069
Encompass_Preferred
Gasket, Sanitary, 2", PTFE/Elastomer Core. Gasket grafts a PTFE envelope to an EPDM elastomer core. Gasket will not creep or cold flow overtime so it maintains the integrity of its seal. Temperature range -73 to 177¿ C.
Catalog No. 50-131-1069 Supplier Chemglass Life Sciences Supplier No. CG152G20
Gasket, Sanitary, 2", PTFE/Elastomer Core. Gasket grafts a PTFE envelope to an EPDM elastomer core. Gasket will not creep or cold flow overtime so it maintains the integrity of its seal. Temperature range -73 to 177¿ C.