Learn More
Chemglass Life Sciences Gasket, Sanitary, 3/4", PTFE/Elastomer Core
SDP
Catalog No. 501311067
Encompass_Preferred
Gasket, Sanitary, 3/4", PTFE/Elastomer Core. Gasket grafts a PTFE envelope to an EPDM elastomer core. Gasket will not creep or cold flow overtime so it maintains the integrity of its seal. Temperature range -73 to 177¿ C.
Catalog No. 50-131-1067 Supplier Chemglass Life Sciences Supplier No. CG152G07
Gasket, Sanitary, 3/4", PTFE/Elastomer Core. Gasket grafts a PTFE envelope to an EPDM elastomer core. Gasket will not creep or cold flow overtime so it maintains the integrity of its seal. Temperature range -73 to 177¿ C.