Learn More
Grainger Gasket Sealant 16 fl oz Brown
Catalog No. 502767896
Encompass_Preferred
The Permatex Gasket Sealant Form-A-Gasket is a high-quality anaerobic gasket maker that seals flanges and fittings. It is formulated to provide resistance to ATF, coolant, oil, and other shop fluids. This gray liquid gasket maker cures when confined in the absence of air between close fitting metal surfaces. It resists aging, weathering, vibration, moisture, fuels, and shop fluids. The product comes in a 2 oz tube, has a temperature range of -65F to 300F when cured, and offers high temperature resistance. It is designed for use on rigid machined flanges with less than .001-inch gap.
Catalog No. 50-276-7896 Supplier Grainger Supplier No. 16X924
The Permatex Gasket Sealant Form-A-Gasket is a high-quality anaerobic gasket maker that seals flanges and fittings. It is formulated to provide resistance to ATF, coolant, oil, and other shop fluids. This gray liquid gasket maker cures when confined in the absence of air between close fitting metal surfaces. It resists aging, weathering, vibration, moisture, fuels, and shop fluids. The product comes in a 2 oz tube, has a temperature range of -65F to 300F when cured, and offers high temperature resistance. It is designed for use on rigid machined flanges with less than .001-inch gap.