Learn More
Grainger Gasket Sheet 12 x 12 In PTFE White
Catalog No. 502764117
Encompass_Preferred
The Grainger 16T698 PTFE Gasket Sheet is a high-quality gasket material made from 100% virgin PTFE. This white gasket sheet has excellent chemical resistance and can withstand temperatures from -450°F to 500°F. It has a tensile strength of 2800 psi and compressibility of 10-16%. This gasket sheet comes in a 12" x 12" size with a thickness of 1/16" and meets military specification MIL-G-24716. It can be used for applications requiring resistance to chemicals, oils, fuels, acids, and solvents. The Grainger 16T698 is an ideal gasket material for use in valves, pumps, compressors, and other equipment requiring a reliable PTFE gasket seal.
Catalog No. 50-276-4117 Supplier Grainger Supplier No. 16T698
The Grainger 16T698 PTFE Gasket Sheet is a high-quality gasket material made from 100% virgin PTFE. This white gasket sheet has excellent chemical resistance and can withstand temperatures from -450°F to 500°F. It has a tensile strength of 2800 psi and compressibility of 10-16%. This gasket sheet comes in a 12" x 12" size with a thickness of 1/16" and meets military specification MIL-G-24716. It can be used for applications requiring resistance to chemicals, oils, fuels, acids, and solvents. The Grainger 16T698 is an ideal gasket material for use in valves, pumps, compressors, and other equipment requiring a reliable PTFE gasket seal.