Learn More
Grainger Gasket Sheet 30 x30 In PTFE w/Glass Blue
Catalog No. 502764635
Encompass_Preferred
The Grainger Approved Gasket Sheet is a high quality gasket material made of neoprene rubber with a pressure sensitive adhesive backing. It has excellent resistance to water, oils, chemicals, vibration, shock, weathering and aging. This gasket sheet comes in a 12" x 12" size, has a thickness of 1/16", and features a maximum temperature rating of 200°F. The pressure sensitive adhesive provides reliable sealing and bonding. This versatile gasket sheet has many potential applications including use as a gasket or seal for pumps, valves, covers, access doors, inspection plates, and more. It conforms well to irregular surfaces. The Grainger item number for this product is 16T704.
Catalog No. 50-276-4635 Supplier Grainger Supplier No. 16T704
The Grainger Approved Gasket Sheet is a high quality gasket material made of neoprene rubber with a pressure sensitive adhesive backing. It has excellent resistance to water, oils, chemicals, vibration, shock, weathering and aging. This gasket sheet comes in a 12" x 12" size, has a thickness of 1/16", and features a maximum temperature rating of 200°F. The pressure sensitive adhesive provides reliable sealing and bonding. This versatile gasket sheet has many potential applications including use as a gasket or seal for pumps, valves, covers, access doors, inspection plates, and more. It conforms well to irregular surfaces. The Grainger item number for this product is 16T704.