Learn More
Helix Medical Inc GASKET SIL 0.5 ; 25 PER BAG
Catalog No. NC1295927
Encompass_Preferred
Biomedical/pharmaceutical grade, platinum-cured silicone clamp gaskets promote crevice-free and protrusion-free processing. The gaskets offer exceptional biocompatibility and contain no contaminants or leachables. May be sterilized by autoclave, ethylene oxide, or gamma radiation.
Catalog No. NC1295927 Supplier Helix Medical Inc Supplier No. 7040005
Biomedical/pharmaceutical grade, platinum-cured silicone clamp gaskets promote crevice-free and protrusion-free processing. The gaskets offer exceptional biocompatibility and contain no contaminants or leachables. May be sterilized by autoclave, ethylene oxide, or gamma radiation.