Learn More
Grainger Gasoline Engine 4 Cycle 7 HP
Catalog No. 502767895
Encompass_Preferred
The Grainger Approved Gasoline Engine 4-Cycle 7 HP is a single cylinder, air-cooled engine with a recoil starter. It has a displacement of 196cc and puts out 7 HP at 3600 RPM. This engine uses regular unleaded gasoline and has a fuel capacity of 0.95 gallons. It comes with a low oil shutdown to prevent damage. The engine has a 3/4" x 2-7/16" crankshaft and includes a muffler. It weighs approximately 39 lbs. This engine is commonly used for pressure washers, log splitters, cement mixers, and go karts. It meets CARB emission standards.
Catalog No. 50-276-7895 Supplier Grainger Supplier No. 16X887
The Grainger Approved Gasoline Engine 4-Cycle 7 HP is a single cylinder, air-cooled engine with a recoil starter. It has a displacement of 196cc and puts out 7 HP at 3600 RPM. This engine uses regular unleaded gasoline and has a fuel capacity of 0.95 gallons. It comes with a low oil shutdown to prevent damage. The engine has a 3/4" x 2-7/16" crankshaft and includes a muffler. It weighs approximately 39 lbs. This engine is commonly used for pressure washers, log splitters, cement mixers, and go karts. It meets CARB emission standards.