Learn More
Gasoline in Water, PT Proficiency Testing Material, Concentrate for dilution to 100 mL, MilliporeSigma™ Supelco™
Catalog No. 111017318 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
2 mL
Catalog No. Quantity
11-101-7318 2 mL
1 options
Catalog No. 11-101-7318 Supplier MilliporeSigma Supelco Supplier No. PE17982ML
This proficiency testing sample is produced in accordance with ISO/IEC 17043:2010. All information regarding the use of this material can be found in the reporting packet supplied for each sample.
This ampule contains a concentrate of a blended gasoline from three different sources in methanol. This PT sample is packaged as a 1.5 mL concentrate. The solvent for this sample is methanol. The PT sample is not preserved.Sample preparationFill a 100-mL volumetric flask to volume with carbon-filtered reagent-grade water at room temperature. Open ampule PE1798-2ML (be careful to avoid cuts on glass) and immediately withdraw 20 muL. Quickly inject the 20 muL aliquot into the reagent water in the expanded area of the volumetric flask. Mix the sample by inverting the flask three times only (excessive shaking results in loss of volatiles) and analyze immediately
TRUSTED_SUSTAINABILITY
Specifications
Packaging Ampule
Quantity 2 mL
Concentration or Composition (by Analyte or Components) Gasoline range organics (GRO), C5-C10 400-4000 μg/L