Learn More
Weber Scientific GASPAK EZ CO2 SACHETS, PK/20
SDP
Catalog No. NC0397617
Encompass
For the BD Diagnostics Gas Generating Container System. GasPak EZ CO2 Container System Sachets. Pack of 20.
Catalog No. NC0397617 Supplier Weber Scientific Supplier No. 309855
For the BD Diagnostics Gas Generating Container System. GasPak EZ CO2 Container System Sachets. Pack of 20.