Learn More
Fritsch Milling And Sizing Inc Gassing Lid Cpl. Hard.Stain.Steel, P-5 Pl
SDP
Catalog No. 501534586
Encompass
Gassing lid P-5 pl with valves and seal ring made of hardened, stainless steel with steel casing
For all grinding bowls premium line 500 ml, 250 ml, 150 ml volume made of agate for PULVERISETTE 5 premium line.
By using this special lid, the grinding bowl can be used for grinding in inert gas and for mechanical alloying - quickly and safely. Gassing lids with Swagelok valves are available on request.
Catalog No. 50-153-4586 Supplier Fritsch Milling And Sizing Inc Supplier No. 50.6557.00
Gassing lid P-5 pl with valves and seal ring made of hardened, stainless steel with steel casing
For all grinding bowls premium line 500 ml, 250 ml, 150 ml volume made of agate for PULVERISETTE 5 premium line.
By using this special lid, the grinding bowl can be used for grinding in inert gas and for mechanical alloying - quickly and safely. Gassing lids with Swagelok valves are available on request.