Learn More
Fritsch Milling And Sizing Inc Gassing Lid Cpl. Agate Premium Line 125-420 Ml
SDP
Catalog No. 501534580
Encompass
Gassing lid cpl. P-5 pl with valves and seal ring made of agate with steel casing
For all grinding bowls premium line 420 ml, 250 ml, 125 ml volume made of agate for PULVERISETTE 5 premium line.
By using this special lid, the grinding bowl can be used for grinding in inert gas and for mechanical alloying - quickly and safely. Gassing lids with Swagelok valves are available on request.
Catalog No. 50-153-4580 Supplier Fritsch Milling And Sizing Inc Supplier No. 50.6407.00
Add to Cart
Add to Cart
Request a Quote
Gassing lid cpl. P-5 pl with valves and seal ring made of agate with steel casing
For all grinding bowls premium line 420 ml, 250 ml, 125 ml volume made of agate for PULVERISETTE 5 premium line.
By using this special lid, the grinding bowl can be used for grinding in inert gas and for mechanical alloying - quickly and safely. Gassing lids with Swagelok valves are available on request.