Learn More
Fritsch Milling And Sizing Inc Gassing Lid Sintered Corundum P-7 Premium Line
SDP
Catalog No. 501534636
Encompass
Sintered corundum gassing lid with valves and seal ring for all grinding bowls premium line 80 ml, 45 ml, 20 ml volume for PULVERISETTE 7 premium line.
By using this special lid, the grinding bowl can be used for grinding in inert gas and for mechanical alloying - quickly and safely.
Gassing lids with Swagelok valves are available on request.
Catalog No. 50-153-4636 Supplier Fritsch Milling And Sizing Inc Supplier No. 50.9637.00
Sintered corundum gassing lid with valves and seal ring for all grinding bowls premium line 80 ml, 45 ml, 20 ml volume for PULVERISETTE 7 premium line.
By using this special lid, the grinding bowl can be used for grinding in inert gas and for mechanical alloying - quickly and safely.
Gassing lids with Swagelok valves are available on request.