Learn More
Fritsch Milling And Sizing Inc Gassing Lid with 2 Valves for 80 Ml Zirconium Oxide, Cl
SDP
Catalog No. 501534617
Encompass
Gassing lid 80 ml cl zirconium oxide with 2 valves and seal ring.
Through the use of this special lid (incl. seal ring) on a zirconium oxide grinding bowl with 80 ml volume, the grinding bowl can be used for grinding with inert gas and mechanical alloying.
Two valves allow for easy and safe filling of the bowls with inert gas, while they are firmly clamped in the mill. A special additional Lock-System is required for gas-tight removal and transportation: order no. 90.1400.00.
Gassing lids with Swagelok valves are available on request.
Catalog No. 50-153-4617 Supplier Fritsch Milling And Sizing Inc Supplier No. 50.8840.00
Gassing lid 80 ml cl zirconium oxide with 2 valves and seal ring.
Through the use of this special lid (incl. seal ring) on a zirconium oxide grinding bowl with 80 ml volume, the grinding bowl can be used for grinding with inert gas and mechanical alloying.
Two valves allow for easy and safe filling of the bowls with inert gas, while they are firmly clamped in the mill. A special additional Lock-System is required for gas-tight removal and transportation: order no. 90.1400.00.
Gassing lids with Swagelok valves are available on request.