Learn More
Fritsch Milling And Sizing Inc Gassing Lid with Valves Wc+Co P-7 Premium Line
SDP
Catalog No. 501534638
Encompass
Hardmetal tungsten carbide gassing lid with valves and seal ring for all grinding bowls premium line 80 ml, 45 ml, 20 ml volume for PULVERISETTE 7 premium line.
By using this special lid, the grinding bowl can be used for grinding in inert gas and for mechanical alloying - quickly and safely.
Gassing lids with Swagelok valves are available on request.
Catalog No. 50-153-4638 Supplier Fritsch Milling And Sizing Inc Supplier No. 50.9647.00
Add to Cart
Add to Cart
Request a Quote
Hardmetal tungsten carbide gassing lid with valves and seal ring for all grinding bowls premium line 80 ml, 45 ml, 20 ml volume for PULVERISETTE 7 premium line.
By using this special lid, the grinding bowl can be used for grinding in inert gas and for mechanical alloying - quickly and safely.
Gassing lids with Swagelok valves are available on request.