Learn More
MSC Gast 0523101QG588NDX 1/4 hp Rotary Vane Vaccum Pump 115/230 Volts, 4.5 CFM, Single Phase, 14" Long x 5.62" Wide x 5.92" High
Catalog No. NC0999986
Encompass
Gast 0523101QG588NDX 1/4 hp Rotary Vane Vaccum Pump 115/230 Volts, 4.5 CFM, Single Phase, 14" Long x 5.62" Wide x 5.92" High
Catalog No. NC0999986 Supplier MSC Supplier No. 80058548
Gast 0523101QG588NDX 1/4 hp Rotary Vane Vaccum Pump 115/230 Volts, 4.5 CFM, Single Phase, 14" Long x 5.62" Wide x 5.92" High