Learn More
Cole-Parmer Gast DOA-P701-AA Oilless Air Compressor, Diaphragm compressor pump, 1.1 cfm, 115 VAC
Catalog No. NC1520422 Shop All Cole-Parmer Products
Encompass
Diaphragm Air Compressors offer oil-free, cool air, even at maximum duty—air entering the pump does not go near the motor to pick up heat.
Catalog No. NC1520422 Supplier Cole-Parmer Supplier No. 0705410
Diaphragm Air Compressors offer oil-free, cool air, even at maximum duty—air entering the pump does not go near the motor to pick up heat.