Learn More
Zeflon International, Inc GASTEC DEW 2-10PPM 10PER BOX
SDP
Catalog No. NC0977192
Encompass
#6LLP Pipeline Dew Point Tube 2-10LB/MMCF. Gastec detector tubes are thin glass tubes with calibration scales printed on them which allows you to directly read concentrations of the substances (gases and vapors) to be measured. Each tube contains detecting reagents that are especially sensitive to the target substance and quickly produces a distinct layer color change.
Catalog No. NC0977192 Supplier Zeflon International, Inc Supplier No. GASTEC6LLP
#6LLP Pipeline Dew Point Tube 2-10LB/MMCF. Gastec detector tubes are thin glass tubes with calibration scales printed on them which allows you to directly read concentrations of the substances (gases and vapors) to be measured. Each tube contains detecting reagents that are especially sensitive to the target substance and quickly produces a distinct layer color change.