Learn More
Med Vet International Gastrogard (Omeprazole) Oral Paste For Equine Ulcers, 6.15gm, Each
Catalog No. 502828840
Encompass_Preferred
VET LICENSE NEEDS TO BE PROVIDED IN 3-4 BUSINESS DAYS OR ORDER WILL BE CANCELLED
Omeprazole (37%) oral paste for the treatment and prevention of gastric ulcers in horses and foals. Omeprazole is a gastric acid pump inhibitor that regulates the final step in hydrogen ion production and blocks gastric acid secretion regardless of the stimulus.
Catalog No. 50-282-8840 Supplier Med Vet International Supplier No. RXGASTRO1
VET LICENSE NEEDS TO BE PROVIDED IN 3-4 BUSINESS DAYS OR ORDER WILL BE CANCELLED
Omeprazole (37%) oral paste for the treatment and prevention of gastric ulcers in horses and foals. Omeprazole is a gastric acid pump inhibitor that regulates the final step in hydrogen ion production and blocks gastric acid secretion regardless of the stimulus.