Learn More
Discovery Life Sciences Gastric Ca Act Tx IO PBMC
SDP
Catalog No. 5000804076
Encompass
PBMCs Peripheral Blood Mononuclear Cells are immune cells isolated from whole blood and viably cryopreserved providing a foundational material for immunology studies cell therapy research and functional immune assays
Catalog No. 50-008-04076 Supplier Discovery Life Sciences Supplier No. 120800
PBMCs Peripheral Blood Mononuclear Cells are immune cells isolated from whole blood and viably cryopreserved providing a foundational material for immunology studies cell therapy research and functional immune assays