Learn More
Discovery Life Sciences Gastric Ca Active Tx PBMC
SDP
Catalog No. 5000804144
Encompass
PBMCs Peripheral Blood Mononuclear Cells are immune cells isolated from whole blood and viably cryopreserved providing a foundational material for immunology studies cell therapy research and functional immune assays
Catalog No. 50-008-04144 Supplier Discovery Life Sciences Supplier No. 120790
PBMCs Peripheral Blood Mononuclear Cells are immune cells isolated from whole blood and viably cryopreserved providing a foundational material for immunology studies cell therapy research and functional immune assays