Learn More
CPC Scientific H-Tyr-Ala-Glu-Gly-Thr-Phe-Ile-Ser-Asp-Tyr-Ser-Ile-Ala-Met-Asp-Lys-Ile-Arg-Gln-Gln-Asp-Phe-Val-Asn-Trp-Leu-Leu-Ala-Gln-Lys-NH2 (trifluoroacetate salt) 0.5MG
SDP
Supplier:  CPC Scientific GIPS002A
Encompass
GIP fragment that presents potent insulinotropic activity in the isolated perfused rat pancreas, but greatly inhibits somatostatinotropic activity in the isolated perfused rat stomach. The site responsible for insulinotropic activity lies between 19 and 30 of GIP. ONE-LETTER SEQUENCE: YAEGTFISDYSIAMDKIRQQDFVNWLLAQK-NH2MOLECULAR FORMULA: C162H245N41O47S1MOLECULAR WEIGHT:3551.04STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [134846-93-8], SYNONYMS: Gastric Inhibitory Polypeptide (1-30) amide (porcine), Glucose-Dependent Insulinotropic Polypeptide (1-30) amide (porcine), GIP (1-30) amide (porcine)RESEARCH AREA: DiabetesREFERENCES: W.J.Rossowski et al., Reg. Pep., 39, 9 (1992)
Catalog No. 50-210-9355