Learn More
CPC Scientific H-Tyr-Ala-Glu-Gly-Thr-Phe-Ile-Ser-Asp-Tyr-Ser-Ile-Ala-Met-Asp-Lys-Ile-His-Gln-Gln-Asp-Phe-Val-Asn-Trp-Leu-Leu-Ala-Gln-Lys-Gly-Lys-Lys-Asn-Asp-Trp-Lys-His-Asn-Ile-Thr-Gln-OH 1MG
SDP
Supplier:  CPC Scientific GIPS001B
Encompass
Incretion is an essential regular of insulin secretion and glucose.SEQUENCE: H-Tyr-Ala-Glu-Gly-Thr-Phe-Ile-Ser-Asp-Tyr-Ser-Ile-Ala-Met-Asp-Lys-Ile-His-Gln-Gln-Asp-Phe-Val-Asn-Trp-Leu-Leu-Ala-Gln-Lys-Gly-Lys-Lys-Asn-Asp-Trp-Lys-His-Asn-Ile-Thr-Gln-OHONE-LETTER SEQUENCE: YAEGTFISDYSIAMDKIHQQDFVNWLLAQKGKKNDWKHNITQMOLECULAR FORMULA: C226H338N60O66S1MOLECULAR WEIGHT: 4983.60STORAGE CONDITIONS: -20 5 CCAS REGISTRY NUMBER: [10040-31-1]SYNONYMS: GIP (human), Glucose-Dependent Insulinotropic Polypeptide (human), GIP (1-42) (human), Gastric Inhibitory Polypeptide (human)RESEARCH AREA: DiabetesREFERENCES:A. Moody et al., FEBS Letters, 123, 205 (1981)SKU(s): GIPS-001A, GIPS-001B
Catalog No. 50-210-9354