Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Gastric inhibitory peptide, porcine | 11063-17-5 | 99.1% | 4975.55 | 1 MG
SDP

Catalog No. 5000399517
Encompass_Preferred

Gastric Inhibitory Peptide, porcine is a glucose-dependent insulinotropic polypeptide. It is a 42 amino acid intestinal hormone that plays a crucial role in fat and glucose metabolism. This product is for research use only.

  • Glucose-dependent insulinotropic polypeptide
  • 42 amino acid intestinal hormone
  • Affects fat and glucose metabolism
  • Purity: 99.12%
  • Molecular weight: 4975.55
  • Appearance: Solid, white to light yellow

Catalog No. 50-003-99517 Supplier Medchemexpress LLC Supplier No. HYP35791MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Gastric Inhibitory Peptide, porcine is a glucose-dependent insulinotropic polypeptide. It is a 42 amino acid intestinal hormone that plays a crucial role in fat and glucose metabolism. This product is for research use only.

  • Glucose-dependent insulinotropic polypeptide
  • 42 amino acid intestinal hormone
  • Affects fat and glucose metabolism
  • Purity: 99.12%
  • Molecular weight: 4975.55
  • Appearance: Solid, white to light yellow

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.