Learn More
Medchemexpress LLC Gastric Inhibitory Peptide, porcine | 11063-17-5 | 99.1% | 4975.55 | 10 MG
SDP
Catalog No. 5000399519
Encompass_Preferred
Gastric Inhibitory Peptide, porcine is a glucose-dependent insulinotropic polypeptide, a 42 amino acid intestinal hormone with effects on fat and glucose metabolism. This product is for research use only and not sold to patients.
  • Glucose-dependent insulinotropic polypeptide
  • 42 amino acid intestinal hormone
  • Affects fat and glucose metabolism
  • For research use only
  • Requires sealed storage, away from moisture and light, under nitrogen
Catalog No. 50-003-99519 Supplier Medchemexpress LLC Supplier No. HYP357910MG
Gastric Inhibitory Peptide, porcine is a glucose-dependent insulinotropic polypeptide, a 42 amino acid intestinal hormone with effects on fat and glucose metabolism. This product is for research use only and not sold to patients.
  • Glucose-dependent insulinotropic polypeptide
  • 42 amino acid intestinal hormone
  • Affects fat and glucose metabolism
  • For research use only
  • Requires sealed storage, away from moisture and light, under nitrogen