Learn More
Bachem Gastric Inhibitory Polypeptide (1-30) amide (porcine) Trifluoroacetate
SDP
Supplier:  Bachem 4030659.1000
Encompass_Preferred
Gastric Inhibitory Polypeptide (1-30) amide (porcine) Trifluoroacetate, H-Tyr-Ala-Glu-Gly-Thr-Phe-Ile-Ser-Asp-Tyr-Ser-Ile-Ala-Met-Asp-Lys-Ile-Arg-Gln-Gln-Asp-Phe-Val-Asn-Trp-Leu-Leu-Ala-Gln-Lys-NH2, 134846-93-8, C162H245N41O47S, Mr 3551.04, 1mg. Glucose-Dependent Insulinotropic Polypeptide (1-30) amide (porcine), GIP (1-30) amide (porcine), Gastric Inhibitory Peptide (1-30), amide, porcine. This GIP fragment has potent insulinotropic activity in the isolated, perfused rat pancreas but greatly reduced somatostatinotropic activity in the isolated perfused rat stomach. The site responsible for insulinotropic activity apparently lies between residues 19 and 30 of GIP.
Catalog No. 50-259-779