Learn More
Bachem Gastric Inhibitory Polypeptide (human)
SDP
Supplier:  Bachem 4030658.0500
Encompass_Preferred
Gastric Inhibitory Polypeptide (human), H-Tyr-Ala-Glu-Gly-Thr-Phe-Ile-Ser-Asp-Tyr-Ser-Ile-Ala-Met-Asp-Lys-Ile-His-Gln-Gln-Asp-Phe-Val-Asn-Trp-Leu-Leu-Ala-Gln-Lys-Gly-Lys-Lys-Asn-Asp-Trp-Lys-His-Asn-Ile-Thr-Gln-OH, 100040-31-1, C226H338N60O66S, Mr 4983.60, 0.5mg. Glucose-Dependent Insulinotropic Polypeptide (human), GIP (human), GIP (1-42) (human). Gastric inhibitory polypeptide (GIP), an incretin, represents an essential regulator of insulin secretion and glucose homeostasis.
Catalog No. NC0426652