Learn More
Biomatik Corporation Gastric Inhibitory Polypeptide (GIP) ELISA Kit
Supplier:  Biomatik Corporation EKU12389
Encompass_Preferred
Gastric Inhibitory Polypeptide (GIP) ELISA Kit for detecting GIP in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Universal. Detection Range: 19.5-5,000pg/ml. Sensitivity: 6.9pg/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Target Synonyms: Incretin hormone; Glucose Dependent Insulinotropic Peptide. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-7973