Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Gastric lipase protein, human (HEK293, His, solution) | >95.0% | 100UG
SDP

Catalog No. 5000175507
Encompass_Preferred

Gastric lipase is a recombinant human gastric (triacylglycerol) lipase produced in HEK293 cells and supplied as a His-tagged solution for research use. It catalyzes the hydrolysis of dietary triglycerides into free fatty acids and glycerol and is useful for biochemical assays, digestion studies, and enzyme characterization.

  • Recombinant human enzyme with native-like glycosylation
  • Expressed in HEK293 cells for improved folding and activity
  • C-terminal His tag for detection and purification
  • High purity (>95%) confirmed by reducing SDS-PAGE
  • Supplied as a ready-to-use solution in multiple size formats
  • Observed molecular weight ~48-50 kDa due to glycosylation

Catalog No. 50-001-75507 Supplier Medchemexpress LLC Supplier No. HYP70262A100UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Gastric lipase is a recombinant human gastric (triacylglycerol) lipase produced in HEK293 cells and supplied as a His-tagged solution for research use. It catalyzes the hydrolysis of dietary triglycerides into free fatty acids and glycerol and is useful for biochemical assays, digestion studies, and enzyme characterization.

  • Recombinant human enzyme with native-like glycosylation
  • Expressed in HEK293 cells for improved folding and activity
  • C-terminal His tag for detection and purification
  • High purity (>95%) confirmed by reducing SDS-PAGE
  • Supplied as a ready-to-use solution in multiple size formats
  • Observed molecular weight ~48-50 kDa due to glycosylation

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.