Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Gastric lipase protein, human | >95.0% | 10UG
SDP

Catalog No. 5000176324
Encompass_Preferred

Recombinant human gastric lipase expressed in HEK293 cells and supplied as a His-tagged solution for research use. The enzyme catalyzes hydrolysis of triacylglycerols and is suitable for enzymatic activity assays, biochemical characterization, and reagent validation. Provided as a 10 μg vial with >95% purity; store at -80°C and avoid repeated freeze-thaw cycles.

  • Recombinant human enzyme expressed in mammalian (HEK293) cells.
  • C-terminal 6xHis tag facilitates purification and detection.
  • High purity (>95%) suitable for biochemical and activity assays.
  • Complete amino acid sequence available for identity confirmation.
  • Stable at -80°C; aliquot to minimize freeze-thaw cycles.

Catalog No. 50-001-76324 Supplier Medchemexpress LLC Supplier No. HYP70262A10UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant human gastric lipase expressed in HEK293 cells and supplied as a His-tagged solution for research use. The enzyme catalyzes hydrolysis of triacylglycerols and is suitable for enzymatic activity assays, biochemical characterization, and reagent validation. Provided as a 10 μg vial with >95% purity; store at -80°C and avoid repeated freeze-thaw cycles.

  • Recombinant human enzyme expressed in mammalian (HEK293) cells.
  • C-terminal 6xHis tag facilitates purification and detection.
  • High purity (>95%) suitable for biochemical and activity assays.
  • Complete amino acid sequence available for identity confirmation.
  • Stable at -80°C; aliquot to minimize freeze-thaw cycles.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.