Learn More
Medchemexpress LLC HY-P1097 1mg Medchemexpress, Gastrin-1, human CAS:10047-33-3 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYP10971MG
Encompass_Preferred
Medchemexpress, HY-P1097 1mg Gastrin-1, human CAS:10047-33-3 Gastrin-1, human is the endogenous peptide produced in the stomach, and increases gastric acid secretion via cholecystokinin 2 (CCK2) receptor. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-4154