Learn More
CPC Scientific Glp-Gly-Pro-Trp-Leu-Glu-Glu-Glu-Glu-Glu-Ala-Tyr-Gly-Trp-Met-Asp-Phe-NH2 0.5MG
SDP
Supplier:  CPC Scientific GAST002A
Encompass
An endogenous peptide, mainly found in the gut, that activates gastric acid secretion, delays gastric emptying and increases blood flow in the gastric mucosa.SEQUENCE: Glp-Gly-Pro-Trp-Leu-Glu-Glu-Glu-Glu-Glu-Ala-Tyr-Gly-Trp-Met-Asp-Phe-NH2ONE-LETTER SEQUENCE: Glp-GPWLEEEEEAYGWMDF-NH2MOLECULAR FORMULA: C97H124N20O31S1MOLECULAR WEIGHT: 2098.23STORAGE CONDITIONS: -20 5 CCAS REGISTRY NUMBER: [10047-33-3]SYNONYMS: Gastrin I (human), Little Gastrin (human), Gastrin-17 (human)RESEARCH AREA: GastrointestinalREFERENCES:R.A. Gregory et al., Nature, 209, 583 (1966)
SKU(s): GAST-002A, GAST-002B, GAST-002C
Catalog No. 50-210-9316